Hexane (mixture of isomers) ACS
Supplier: INNOVA CHEMICALS CA
|
Danger
|
INN890077
INN890078
INN890080
CA75816-040CS
415
CAD
CA75816-040
CA75816-042
CA75816-046
Hexane (mixture of isomers) ACS
Hexane (mixture of isomers)
Meets ACS specifications for general use.
Formula:
H₃C(CH₂)₄CH₃ Boiling Pt: 68…70 °C (1013 hPa) Melting Pt: –95 °C Density: 0.66 g/cm³ (20 °C) Flash Pt: –22 °C |
MDL Number:
MFCD02179311 CAS Number: 110-54-3 UN: 1208 ADR: 3,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...