Heptane (mixture of isomers) ACS

Supplier: INNOVA CHEMICALS CA
Danger

INN890073 INN890074 INN890075
CA75816-034EA 375 CAD
CA75816-034 CA75816-036 CA75816-038
Heptane (mixture of isomers) ACS
Heptane (mixture of isomers)

Suitable for organic synthesis, cleaning, and isocratic and preparative LC. Clear, colorless liquid.

Formula: H₃C(CH₂)₅CH₃
MW: 100.2 g/mol
Boiling Pt: 98.4 °C (1013 hPa)
Melting Pt: -90.5
Density: 0.6838 g/cm³ (20 °C)
Flash Pt: –4 °C
MDL Number: MFCD00009544
CAS Number: 142-82-5
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dabde8793eb27ef796098b842bf18b9ced8539b8&vid=d30a8083280d1915a43f51d93c49f9b76eabd579&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR