Ethyl acetate ACS

Supplier: INNOVA CHEMICALS CA
Danger

INN890046 INN890048 INN890049 INN890051
CA75816-014CS 101.93 CAD
CA75816-014 CA75816-016 CA75816-020 CA75816-022 CA75816-026
Ethyl acetate ACS
Ethyl acetate

Meets ACS Specifications for General Use


Synonyms: Acetic acid ethyl ester 

Formula: CH₃COOC₂H₅
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
MDL Number: MFCD00009171
CAS Number: 141-78-6
UN: 1173
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Liquid, clear color


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR