Toluene ACS

Supplier: INNOVA CHEMICALS CA
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

INN890162 INN890163 INN890165
CA75815-788CS 416 CAD
CA75815-788 CA75815-790 CA75815-794
Toluene ACS
Toluene

Suitable for  histology and cytology applications.


Synonyms: Methylbenzene , Phenylmethane 

Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6
Melting Pt: -95
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
MDL Number: MFCD00008512
CAS Number: 108-88-3
UN: 1294
ADR: 3 II
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR