Kerosene, odorless

Supplier: INNOVA CHEMICALS CA
Danger

Synonyms: C13-14 isoparaffin, Light distillate

INN890124 INN890125
CA75815-782CS 250 CAD
CA75815-782 CA75815-784
Kerosene, odorless
Kerosene - CAS 64742-47-8
Boiling Pt: 151.1…301 °C (304…574 °F)
Melting Pt: –17.8 °C / 0 °F
Flash Pt: 71.1 °C (160 °F)
CAS Number: 64742-47-8
UN: 1223
ADR: 3,III

Order Now

Specification Test Results

Appearance/color Liquid, light yellow


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR