Methanol ACS

Supplier: INNOVA CHEMICALS CA
Danger

INN890107 INN890108 INN890109 INN890110 INN890111
CA75815-766CS 115 CAD
CA75815-766 CA75815-768 CA75815-770 CA75815-772 CA75815-774
Methanol ACS
Methanol

Synonyms: Carbinol , Hydroxy methane , Methyl alcohol , Wood alcohol 

Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Liquid, clear color


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR