2-Propanol ACS

Supplier: INNOVA CHEMICALS CA
Danger

INN890090 INN890091 INN890092 INN890094
CA75816-052CS 450 CAD
CA75816-052 CA75816-054 CA75816-056 CA75816-060
2-Propanol ACS
2-Propanol

Synonyms: 2-Hydroxy propane, Dimethyl carbinol, IPA, Isopropanol, Isopropyl Rubbing Alcohol, Isopropyl alcohol, Propan-2-ol

Suitable for histology and cytology applications. Soluble in water. Meets ACS Specifications for general use.

Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Liquid, clear color


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR