Acetone ACS

Supplier: INNOVA CHEMICALS CA
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

INN890009 INN890005 INN890006 INN890007
CA75815-758EA 3116 CAD
CA75815-758 CA75816-062 CA75816-064 CA75816-066
Acetone ACS
Acetone

Suitable for histology, organic synthesis, cleaning, isocratic, preparative LC.and cytology applications.Soluble in water

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56 °C (1013 hPa)
Melting Pt: –94 °C
Density: 0.791 g/cm³ (25 °C) (77 °F)
Flash Pt: –17 °C
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Liquid, clear color

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR