Xylene (mixture of isomers) ACS

Supplier: INNOVA CHEMICALS CA
Danger

Synonyms: Dimethylbenzene

INN890178 INN890179 INN890180 INN890182
CA75815-708CS 529 CAD
CA75815-708 CA75815-710 CA75815-712 CA75815-716
Xylene (mixture of isomers) ACS
Xylene (mixture of isomers)

Meets ACS Specifications, Meets Reagent Specifications

Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 140
Density: 0.86…0.88 g/cm³ (20 °C)
Flash Pt: 25 °C
MDL Number: MFCD00077264
CAS Number: 1330-20-7
UN: 1307
ADR: 3 III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR