Helix® 1092 Gloves, HexArmor
Supplier: HexArmor
1092-XXS(5) 1092-XS(6) 1092-S(7) 1092-M(8) 1092-L(9) 1092-XL(10) 1092-XXL(11) 1092-XXXL(12)
76208-000PR 15.97 CAD
76208-000 76208-002 76208-004 76208-014 76208-016 76208-018 76208-020 76208-022
Helix® 1092 Gloves, HexArmor
Gloves
The Helix® Series 1092 features a high-performance nylon blend shell and HexArmor’s proprietary Safe Finger Release (SFR) technology for applications that involve drills or moving parts.
  • Knit: High-performance 15-gauge nylon blend shell
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Safe Finger Release (SFR) technology for applications that involve drilling or moving parts
A silicone-free, flexible foam nitrile palm coating provides workers with superior grip and abrasion resistance.