DuPont™ Tyvek® 400 Pants
Supplier: DuPont
TY350SWHSM005000 TY350SWHMD005000 TY350SWHLG005000 TY350SWHXL005000 TY350SWH2X005000 TY350SWH3X005000 TY350SWH4X005000 TY350SWH5X005000 TY350SWH6X005000 TY350SWH7X005000
CA10010-111CS 1443.87 CAD
CA10010-111 CA10010-113 CA10010-115 CA10010-117 CA10010-119 CA10010-121 CA10010-123 10016-944 10016-946 10016-948
DuPont™ Tyvek® 400 Pants
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Open ankles
  • Elastic waist
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous filmMore Product Information