Hercules® 400R6E Gloves, HexArmor
Supplier: HexArmor
400R6E-XL(10) 400R6E-XXL(11) 400R6E-M(8) 400R6E-L(9)
76208-030PR 825.16 CAD
76208-030 76208-032 76208-470 76208-472
Hercules® 400R6E Gloves, HexArmor
The Hercules® 400R6E are no stranger to dangerous applications such as handling razor wire or protecting from animal bites. Gauntlet design and pre-curved shape for maximum comfort and protection.
  • SuperFabric®* brand material palm provides ANSI/ISEA cut level A9 resistance for some of the highest cut protection available in the industry (over entire hand)
  • Innovative industrial puncture protection over the entire hand from wires, metal, wood, glass, and other puncture related hazards
  • Gauntlet design and pre-curved shape for maximum comfort and dexterity
  • Specialized silicon gripping surface on palm
  • Launderable
The Hercules® 400R6E combines industrial puncture protection and an ANSI/ISEA A9 score by layering HexArmor’s exclusively-licensed SuperFabric® brand material. Full hand coverage from wires, metal, wood, glass, and razorwire.