Bandit® 5042 Gloves, HexArmor
Supplier: Hexarmor
BAND-IT®
5042-M(8) 5042-XL(10) 5042-XXL(11) 5042-L(9) 5042-S(7)
76208-098PR 182.76 CAD
76208-098 76208-050 76208-052 76208-046 76208-048
Bandit® 5042 Gloves, HexArmor
Gloves
The Bandit® 5042 is a heavy-duty leather work glove made in the USA with U.S. and imported components. With an interior layer of high-performance Corecept® technology, the 5042 provides industrial puncture protection against the most hazardous sharps and splinters found in the lumber industry.
  • High-performance Corecept® technology provides industrial puncture protection (interior layer)
  • ANSI/ISEA level A5 cut resistance in enhanced areas
  • Premium cowhide leather palm provides a traditional style of comfort
  • Reinforced seams
  • Grip in dry or light oil situations
  • Canvas safety cuff
Bandit® 5042 gloves feature heavy-duty cowhide leather lined with high-performance Corecept® technology for industrial puncture protection. Popular in lumber industries and made in the USA, this glove offers ANSI/ISEA A5 cut resistance in strategic areas.