PointGuard® Ultra 4041 Gloves, HexArmor
Supplier: HexArmor
4041-XXS(5) 4041-XS(6) 4041-S(7) 4041-M(8) 4041-L(9) 4041-XL(10) 4041-XXL(11)
76208-100PR 306.88 CAD
76208-100 76208-102 76208-104 76208-114 76208-116 76208-118 76208-120
PointGuard® Ultra 4041 Gloves, HexArmor
Gloves
The PointGuard® Ultra 4041 is the only mechanic’s style glove to have both cut and needle-resistance, giving end users superior protection, a performance advantage to keep them safer on the job. Designed with specialized silicone gripping surface on palm.
  • Highest needlestick resistance available provided by the layering of SuperFabric®* brand material
  • Tested using actual 25 gauge needles against ANSI/ISEA 105 standard
  • Back-of-hand knuckle padding for incidental bumps and impacts
  • Neoprene cuff with Velcro® closure for secure fit
  • Silicone pattern palm grip for enhanced durability and grip
  • Form-fitting design for increased dexterity and comfort
  • Launderable
Rated ANSI/ISEA A9 for cut and Level 5 in needlestick, this glove is popular in law enforcement and public safety settings.