NXT™ 10-302 Gloves, HexArmor
Supplier: HexArmor
10-302-XXS(5) 10-302-XXL(11) 10-302-XXXL(12)
76207-956EA 63.82 CAD
76207-956 76207-958 76207-960 76207-962 76207-964 76207-966 76207-968 76207-970
NXT™ 10-302 Gloves, HexArmor
Gloves
The NXT™ 10-302 is an ambidextrous food service glove with SuperFabric® brand materials for coverage that targets primary knife strike areas, on top of a performance shell that makes it a true revolution in hand protection. Cool, comfortable, and provides great dexterity.
  • 3-finger cut resistant SuperFabric®* brand material coverage targets primary knife strike areas
  • High dexterity CoreTek® high performance shell keeps hands cool and comfortable during work
  • Meets European food migration testing (EN 1186:2002 SI 1998 No 1376)
  • Ambidextrous
  • Launderable/sanitizeable (can be washed in a commercial dishwasher)
The NXT™ ambidextrous glove with SuperFabric® brand material armor plating on top of a CoreTek® cut resistance performance shell provides high cut-resistance and dexterity while remaining cool and comfortable in any food production environment.