AG10009S Protective Arm Guards, HexArmor
Supplier: HexArmor
AG10009S-XS(6) AG10009S-S(7) AG10009S-M(8) AG10009S-L(9) AG10009S-XL(10) AG10009S-XXL(11) AG10009S-XXXL(12)
76207-942EA 135.56 CAD
76207-942 76207-944 76207-946 76207-948 76207-950 76207-952 76207-954
AG10009S Protective Arm Guards, HexArmor
Gloves
The AG10009S offers industrial puncture protection over the entire forearm with a spandex wrist insert and thumbhole. No more snags, punctures, abrasions, or cuts to the forearm with this unique product.
  • Made with SuperFabric® brand material for some of the highest cut-resistance available on the market, ANSI/ISEA level A7 and CE level 5
  • Innovative industrial puncture protection from wires, metal, wood, and glass over the entire forearm
  • Won’t fall down like knit sleeves, lightweight and cooler than knit or leather alternatives
  • Lasts 13 times longer than competitive sleeves
  • Spandex wrist insert with thumbhole
  • Launderable
Industrial puncture forearm protection with spandex wrist insert and thumbhole that won’t slide like other knit armguard materials. Eliminate snags, punctures, abrasions, and cuts with this SuperFabric® brand material armguard made in the USA.