Chrome Series® 4027 Gloves, HexArmor
Supplier: Hexarmor
4027-XXXL(12) 4027-L(9) 4027-S(7) 4027-XL(10) 4027-M(8) 4027-XXL(11)
76207-938PR 169.9 CAD
76207-938 76207-936 76207-934 76207-932 76207-930 76207-940
Chrome Series® 4027 Gloves, HexArmor
Gloves
The Chrome Series® 4027 features a SuperFabric® brand material palm that provides ANSI/ISEA level A8 cut resistance giving over 5000 grams of protection, 20 times the amount offered by a standard leather glove.
  • SuperFabric®* brand material palm provides ANSI/ISEA level A8 cut resistance for some of the highest cut-resistance available in the industry
  • Superior back of hand impact protection system utilizes an advanced design to dissipate forceful blows over a large area
  • Durable TP-X® palm utilizes the highest-level abrasion resistance while maintaining an oil-resistant grip
  • Hi-vis color and reflective tape on back-of-hand
  • Elastic cuff with Velcro® closure for a secure fit
  • Launderable
Chrome Series® 4027 features SuperFabric® brand material in the palm providing ANSI/ISEA level A8 cut resistance and over 5000 grams of protection (20 times the amount found in a standard leather glove). This glove also features a durable TP-X® palm and full back of hand impact protection system.