Econo™ Jeweler Style Forceps, Floor Grade, Sklar®
Supplier: Sklar
21-915 21-817 21-819
76192-706BX 289.57 CAD
76192-706 76192-820 76192-822
Econo™ Jeweler Style Forceps, Floor Grade, Sklar®
Forceps
Econo™ Jeweler Style Forceps are grasping forceps used in microsurgical procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Econo™ Jeweler Style Forceps have fine or micro-fine tips that can handle extremely fine tissues and materials that standard instruments may damage. They are multipurpose thumb forceps with flat handles available with various tip styles. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.