TC Corwin Wire Twister, OR Grade, Sklar
Supplier: Sklar
21-9050 21-9051 21-9052
76192-852EA 2484.28 CAD
76192-852 76192-854 76192-856
TC Corwin Wire Twister, OR Grade, Sklar
Pliers
The Sklar® Tungsten Carbide Corwin Wire Twister is a ratched, three ring instrument used in a variety of procedures where wire is used.
  • Industry standard
  • Made of tungsten carbide/stainless steel
  • Tip allows for a firm grip
  • These are identified by gold handles
These are used to manipulate wire to stabilize bones. Sklar® permanently bonds tungsten carbide inserts to the instrument, ensuring sharpness for a longer period of time, and maximizing control and precision.
Order Now
SPECIFICATIONS
Environmentally Preferable