Gregory Stay Suture Clamp Forceps, OR Grade, Sklar®
Supplier: Sklar
24-3010 24-3011 24-3012 24-3013
76192-982EA 922.43 CAD
76192-982 76192-984 76192-986 76192-988
Gregory Stay Suture Clamp Forceps, OR Grade, Sklar®
Forceps
Sklar® Gregory Stay Suture Clamp Forceps are small, lightweight forceps with smooth jaws that are used during microsurgical procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The small clamps allow sutures to be temporarily placed to hold the area that is being operated on. The instrument comes in a variety of lengths and curvatures.