Langenbeck Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8081 21-8083 21-8085 21-8069
76192-676EA 521.07 CAD
76192-676 76192-678 76192-680 76192-730 76192-732 76192-734
Langenbeck Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Langenbeck needle holder is used for delicate and refined suturing.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Langenbeck needle holder features plier handles, but has no spring action and no locking mechanism. This instrument is often chosen by users who prefer a quick release of the needle when suturing.