TC Arruga Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-3100 21-3101
76192-350EA 536.2 CAD
76192-350 76192-352
TC Arruga Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® TC Arruga Needle Holder is primarily used in ophthalmologic procedures such as eyelid surgery.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Sklar® TC Arruga Needle Holder is a single spring instrument with a ratchet locking mechanism at the base of the rectangular plier handle. This product is straight with cross-serrated tungsten carbide tips. Sklar® permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.