TC Senning Needle Holder, OR Grade, Sklar®
Supplier: Sklar
20-2311 20-2313 20-2315
76192-510EA 1159.19 CAD
76192-510 76192-512 76192-514
TC Senning Needle Holder, OR Grade, Sklar®
Forceps
The Sklar® Sennning TC Needle Holder is commonly used in vascular and plastic surgery procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The Sennning TC Needle Holder is a ratcheted, finger ring instrument with short jaws and shanks that bend inward slightly, right above the ratchet, similar to those of a Sarot needle holder. This slight bend creates a longer, narrow section of shanks providing access to tight surgical areas.