DuPont™ Tychem® 4000 Coveralls with Standard Hood, Taped Seams
Supplier: DuPont
Tychem®
SL127TWH7X000600 SL127TWH3X000600 SL127TWHXL00 SL127TWHXL00 SL127TWH4X00 SL127TWH3X00 SL127TWHMD00 SL127TWH4X00 SL127TWH6X000600 SL127TWHLG000600 SL127TWHMD00 SL127TWH5X000600 SL127TWH2X00 SL127TWH2X00
CA13451-630CS 1883.07 CAD
CA13451-630 13451-622 13451-632 13451-626 CA13451-632 CA13451-622 13451-628 CA13451-628 CA13451-626 89207-858 10016-496 10016-498 89207-854 10016-500
DuPont™ Tychem® 4000 Coveralls with Standard Hood, Taped Seams
Disposable Garments
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
  • Standard fit hood
  • Elastic wrists and ankles
  • Storm flap with adhesive closure
  • Taped seams
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Coveralls feature an attached hood with elastic around face opening and elastic openings at the wrists and ankles for a tighter fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper.
Certifications: Thickness meets ASTM D1777.