DuPont™ Tychem® 4000 Coveralls with Laydown Collar and Elastic Wrists and Ankles, Taped Seams
Supplier: DuPont
Tychem®
SL125TWH2X00 SL125TWHMD00 SL125TWHLG00 SL125TWH3X00 SL125TWHXL00 SL125TWH2X00 SL125TWHMD00 SL125TWH5X000600 SL125TWH4X00 SL125TWH4X00 SL125TWHLG00 SL125TWH3X00 SL125TWHXL00
CA13451-598CS 1569.42 CAD
13451-596 13451-598 CA13451-598 CA13451-596 13451-602 CA13451-602 13451-604 13451-606 CA13451-606 13451-608 CA13451-604 CA13451-608 10016-488
DuPont™ Tychem® 4000 Coveralls with Laydown Collar and Elastic Wrists and Ankles, Taped Seams
Disposable Garments
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
  • Laydown collar
  • Elastic wrists and ankles
  • Storm flap with adhesive closure
  • Taped seams
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications include chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Coveralls feature a laydown collar, and elastic openings at wrists and ankles for a tighter fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper.
Certifications: Thickness meets ASTM D1777.