DuPont™ Tychem® 2000 Coveralls with Standard Hood and Elastic Wrists and Ankles, Taped Seams
Supplier: DuPont
Tychem®
QC127TYL5X000400 QC127TYL2X00 QC127TYLLG00 QC127TYLMD00 QC127TYLLG00 QC127TYLXL00 QC127TYL2X00 QC127TYL4X00 QC127TYL3X00 QC127TYL3X00 QC127TYLXL00 QC127TYLMD00 QC127TYL4X00
CA10010-440CS 1001.39 CAD
10010-440 CA10010-440 CA10010-442 10010-432 10010-442 CA10010-436 10010-436 CA10010-438 10010-434 CA10010-432 CA10010-434 10010-438 10016-424
DuPont™ Tychem® 2000 Coveralls with Standard Hood and Elastic Wrists and Ankles, Taped Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Standard fit hood
  • Elastic wrists and ankles
  • Serged seams
  • Available NAFTA sourced
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Attached hood with elastic around face opening is standard fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Elastic openings at wrists and ankles allow for a tighter fit.
Certifications: Thickness meets ASTM D1777.