DuPont™ Tychem® 2000 Coveralls with Standard Hood and Elastic Wrists and Ankles, Serged Seams
Supplier: DuPont
QC127SYLMD001200 QC127SYLLG001200 QC127SYLXL001200 QC127SYL2X001200 QC127SYL3X001200 QC127SYL4X001200 QC127SYL5X001200 QC127SYL6X001200 QC127SYL7X001200
CA10010-210CS 1034.63 CAD
CA10010-210 CA10010-212 CA10010-214 CA10010-216 CA10010-218 CA10010-220 CA13452-664 10016-408 10016-410 10016-412 10016-414 10016-416 10016-418 10016-420 10016-422
DuPont™ Tychem® 2000 Coveralls with Standard Hood and Elastic Wrists and Ankles, Serged Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Standard fit hood
  • Elastic wrists and ankles
  • Serged seams
  • Available NAFTA sourced
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Multiple interlockingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application Cleanroom,Laboratory