DuPont™ Tychem® 2000 Coveralls with Laydown Collar and Elastic Wrists and Ankles, Taped Seams
Supplier: DuPont
QC125TYLMD000400 QC125TYLLG000400 QC125TYL2X000400 QC125TYL3X000400 QC125TYL4X000400 QC125TYL5X000400
CA10010-418CS 833.76 CAD
CA10010-418 CA10010-420 CA10010-424 CA10010-426 CA10010-428 CA89067-942
DuPont™ Tychem® 2000 Coveralls with Laydown Collar and Elastic Wrists and Ankles, Taped Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Laydown collar
  • Elastic wrists and ankles
  • Storm flap with adhesive closure
  • Taped seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Taped seams provideMore Product Information