DuPont™ Tychem® 2000 Coveralls with Laydown Collar and Elastic Wrists and Ankles, Serged Seams
Supplier: DuPont
Tychem®
QC125SYL4X00 QC125SYLLG00 QC125SYL7X001200 QC125SYLLG00 QC125SYLMD00 QC125SYL3X00 QC125SYL5X00 QC125SYL5X00 QC125SYL2X00 QC125SYLXL00 QC125SYL6X001200 QC125SYL3X00 QC125SYLMD00 QC125SYL4X00 QC125SYLXL00 QC125SYL2X00
CA13452-660CS 1019.02 CAD
13452-658 13452-656 13452-654 13452-652 13452-662 13452-650 13452-660 CA13452-660 CA13452-650 10016-400 CA13452-662 CA13452-652 10016-402 CA13452-654 CA13452-656 CA13452-658
DuPont™ Tychem® 2000 Coveralls with Laydown Collar and Elastic Wrists and Ankles, Serged Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Laydown collar
  • Elastic wrists and ankles
  • Serged seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls feature a laydown collar, elastic wrists, and elastic ankles.
Certifications: Thickness meets ASTM D1777.