DuPont™ Tychem® 2000 Coveralls with Standard Hood and Attached Socks, Taped Seams
Supplier: DuPont
QC122TYLMD000400 QC122TYLLG000400 QC122TYLXL000400 QC122TYL2X000400 QC122TYL3X000400 QC122TYL4X000400 QC122TYL5X000400 QC122TYL6X000400
CA89138-050CS 955.75 CAD
CA89138-050 CA89138-052 CA89138-054 CA89138-056 CA89138-058 CA89138-060 CA89138-062 10016-394
DuPont™ Tychem® 2000 Coveralls with Standard Hood and Attached Socks, Taped Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Standard fit hood
  • Elastic wrists
  • Attached socks
  • Storm flap with adhesive closure
  • Taped seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Attached hood with elastic around face opening is standard fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Elastic openings at wrists allow for a tighter fit. Integrated socks are composed of the same garment material.
Certifications: Thickness meets ASTM D1777.