DuPont™ Tychem® 2000 Coveralls with Standard Hood and Attached Socks, Serged Seams
Supplier: DuPont
QC122SYLMD001200 QC122SYLLG001200 QC122SYLXL001200 QC122SYL2X001200 QC122SYL3X001200 QC122SYL4X001200 QC122SYL5X001200 QC122SYL6X001200 QC122SYL7X001200 QC122SYL8X001200
CA10010-196CS 1120.15 CAD
CA10010-196 CA10010-198 CA10010-200 CA10010-202 CA10010-204 CA10010-206 CA13452-632 10016-388 10016-390 10016-392
DuPont™ Tychem® 2000 Coveralls with Standard Hood and Attached Socks, Serged Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Standard fit hood
  • Elastic wrists
  • Attached socks
  • Serged seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Attached hood with elastic around face opening is standard fit. Elastic openings at wrists allow for a tighter fit. Integrated socks are composed of the same garment material.
Certifications: Thickness meets ASTM D1777.