USP

Supplier: TEKNOVA CA

C2101 C2110 C2113
CA100216-810EA 279.27 CAD
CA100216-810 CA100217-982 CA101414-094
USP
Carbenicillin

Carbenicillin is a semi-synthetic ampicillin analog, and like ampicillin is a beta-lactam antibiotic. Carbenicillin is a member of the carboxypenicillin subgroup of the penicillins.


It is effective against Gram-negative organisms including H. influenzae and E. coli. Carbenicillin is also used in transformed Agrobacter species. Carbenicillin was the first penicillin with significant activity against Pseudomonas species.

Formula: C₁₇H₁₈N₂O₆S
CAS Number: 4697-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR