DURAN®PURE® Bottles, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
818014407 818015403 818016305 818017301 818018606 818019105 818011401 818012406 818013608 818063604 818064403 818065408 818066301 818067306 818068602 818069101 818604403 818605408 818665405 818666307 891081306 891088679 891169109 898862802 818061406 818062402 818606301 818664409
76076-868CS 352.84 CAD
76076-868 76076-870 76076-872 76076-874 76076-876 76076-878 76076-880 76076-882 76076-884 76076-886 76076-888 76076-890 76076-892 76076-894 76076-896 76076-898 76076-900 76076-902 76076-904 76076-906 76076-908 76076-910 76076-912 76202-920 76202-922 76202-924 76202-926 76202-928
DURAN®PURE® Bottles, DWK Life Sciences
Bottles Glass Bottles
Intermediate biologic bottle system with comprehensive qualification and validation package.
  • USP compliant range of bottles from 25 ml to 20 L
  • Chemically inert
  • Wide usage temperature range
  • Excellent gas barrier
  • Low leachable/extractable profile
  • Biocompatibility
Bottles are made from Type 3.3 borosilicate glass. A protective dust cover is applied after the bottle is cooled on the production line.
DURAN PURE Premium Screw Caps are manufactured from fluoropolymer. The caps are colorant- and pigment-free and are suitable for most sterilization methods.
More Product Information