DuPont™ ProClean® Frocks, Front Zipper Closure
Supplier: DuPont
PC271SWHLG00300B PC271SWH2X00300B PC271SWH3X00300B PC271SWH4X00300B PC271SWHSM00300B PC271SWHMD00300B PC271SWHXL00300B
CA13452-546CS 1222.21 CAD
CA13452-546 CA13452-550 CA13452-552 CA13452-554 CA89129-704 CA13452-544 CA13452-548
DuPont™ ProClean® Frocks, Front Zipper Closure
Disposable Garments
ProClean® offers improved non-hazardous light liquid splash protection.
  • Serged seams
  • Bound neck
  • Set sleeve design
  • Covered elastic wrists
  • Front zipper closure
ProClean® is made from a microporous composite fabric. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Covered elastic openings at wrists allows for a tighter fit. Bound neck ensures lower particle shedding. Frocks provide body-length jacket design with front zipper closure for easy donning and doffing.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.