G418 disulfate solution 50 mg/ml, Corning®

Supplier: Corning
Danger

30-234-CR 30-234-CI
CA45000-630EA 266.44 CAD
CA45000-630 CA45001-076
G418 disulfate solution 50 mg/ml, Corning®
G418 disulphate

G418 Sulfate is recommended for use as selection agent in a concentration range of 100-800 µg/ml.


This antibiotic blocks polypeptide synthesis by hindering protein elongation, and inhibits the selection and maintenance of eukaryotic cells stably transfected with the neomycin resistance gene. G418 is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.


Caution: For Research or Further Manufacturing Use Only. Not for use in Diagnostic Procedures.

Formula: C₂₀H₄₄N₄O₁₈S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR