Beebee Crown Scissors, OR Grade, Sklar®
Supplier: Sklar
24-2330 24-2331 24-2332 24-2333
76192-964EA 236.83 CAD
76192-964 76192-966 76192-968 76192-970
Beebee Crown Scissors, OR Grade, Sklar®
Scissors
Sklar® Beebee Crown Scissors are a dental instrument used for cutting a variety of materials when creating temporary crowns.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Beebee Crown Scissors have short, stout jaws that may be either straight or curved, with sharp or blunt blades, useful for cutting heavier material.