Sklar Edge™ Tungsten Carbide Supercut Sklarhone™ Kilner Scissors, OR Grade, Sklar®
Supplier: Sklar
16-3619 16-3620 16-3621
76192-144EA 1177.08 CAD
76192-144 76192-146 76192-148
Sklar Edge™ Tungsten Carbide Supercut Sklarhone™ Kilner Scissors, OR Grade, Sklar®
Scissors
Sklarhone™ Kilner (Ragnell) scissors are used for the dissecting and cutting of delicate tissue during minor procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar Edge™ TC Supercut Sklarhone™ line of scissors provide effortless, precise cutting. These scissors feature one knife-like razor edge and one micro-serrated edge along with the strength and durability of tungsten carbide. Sklar permanently bonds tungsten carbide inserts to the blades, which maximizes control and precision. These scissors are also designed to stay sharp for a significantly longer period of time than standard scissors. This instrument can be identified by its handle of one black finger ring and one gold finger ring.