Sklar Edge™ Tungsten Carbide Supercut Sklarhone™ Iris Scissors, OR Grade, Sklar®
Supplier: Sklar
16-3545 16-3546
76192-078EA 772.81 CAD
76192-078 76192-080
Sklar Edge™ Tungsten Carbide Supercut Sklarhone™ Iris Scissors, OR Grade, Sklar®
Scissors
Sklar Edge™ TC Supercut Sklarhone™ Iris scissors provide effortless, precise cutting.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
These scissors feature one knife-like razor edge and one micro-serrated edge along with the strength and durability of tungsten carbide. They can be used to cut both delicate and heavy tissue with minimal tearing or pulling. Sklar permanently bonds tungsten carbide inserts to the blades, which maximizes control and precision. These scissors are also designed to stay sharp for a significantly longer period of time than standard scissors. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures.