HiLoad Superdex Columns, 75 PG, Cytiva
Supplier: Cytiva
HiLoad™
28-9893-33 28989333 28989334 28-9893-34
CA71002-668EA 13297.68 CAD
CA71002-668 CA71002-666 71002-668 71002-666
HiLoad Superdex Columns, 75 PG, Cytiva
Chromatography Columns
Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component.
  • High resolution with short run times and good recovery
  • High capacity at high flow rates
  • Separations can be scaled up to production levels
  • Prepacked for convenience and reproducibility
  • ¹/₁₆" fittings for convenient connections to ÄKTA design systems
Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr <10000 (Superdex 30 pg), Mr 3000 to 70000 (Superdex 75 pg), and Mr 10000 600000 (Superdex 200 pg). The medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions.
Delivery information: Includes dismantling tool, support screen, net ring (10 μm), O-ring, stop plugs and instructions.