PIPETMAN® Expert Sterilized MEGAPACK Tips, Gilson®
Supplier: GILSON, INC.
F1733072 F1732072 F1733071 F1732071
76178-316CS 647.32 CAD
76178-316 76178-384 76180-442 76180-444
PIPETMAN® Expert Sterilized MEGAPACK Tips, Gilson®
Pipette Tips
Universal fit PIPETMAN® EXPERT Tips are designed to fit leading pipette brands, allowing you to stock one brand of tips saving time, money, and bench space. The tips are produced using state-of-the-art manufacturing techniques assuring lot-to-lot consistency and high performance. Volume graduations make it easy to visually check the volume aspirated. Color-coded packaging corresponds to industry standards for color matching to respective Gilson pipette models.
  • Universal Fit
  • Free of detectable contaminants
  • convenient, economical packaging
Universal fit EXPERT Tips fit leading pipette brands eliminating the need to stock multiple tip brands. EXPERT Tips are manufactured in a sterile, clean-room environment and are certified free of detectable human DNA, RNases, DNases, PCR inhibitors, and pyrogens.
EXPERT Tips come in a wide range of packaging options to fit your needs. From bulk tips to convenient racked tips, they are available sterilized or in autoclavable packaging.
Packaging: Packaging features 192 tips per layer, stacked 5 layers high.