PIPETMAN® Expert TowerPack™ Tips, Gilson®
Supplier: GILSON, INC.
F1735061 F1736061 F1736052 F1732061 F1732052 F1736051 F1732051 F1735052 F1733061 F1733052 F1735051 F1733051
76178-314PK 164.98 CAD
76178-314 76178-322 76178-324 76178-340 76178-382 76178-446 76178-448 76180-038 76180-370 76180-372 76180-452 76180-454
PIPETMAN® Expert TowerPack™ Tips, Gilson®
Pipette Tips
Universal fit PIPETMAN® EXPERT Tips are designed to fit leading pipette brands, allowing you to stock one brand of tips saving time, money, and bench space. The tips are produced using state-of-the-art manufacturing techniques assuring lot-to-lot consistency and high performance. Volume graduations make it easy to visually check the volume aspirated. Color-coded packaging corresponds to industry standards for color matching to respective Gilson pipette models.
  • Universal Fit
  • Free of detectable contaminants
  • convenient, economical packaging
Universal fitMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable