PIPETMAN® Empty Tip Racks, Gilson®
Supplier: GILSON, INC.
F1734021 F1735021 F1732021 F1733021 F1732022 F1735022 F1733022 F1734022
76178-380EA 23.96 CAD
76178-380 76178-462 76178-464 76178-466 76178-478 76178-480 76178-488 76178-490
PIPETMAN® Empty Tip Racks, Gilson®
Pipette Tip Boxes and Racks
Purchase in bulk and save money by simply reloading these tips directly into the PIPETMAN® EXPERT Empty Racks.
  • Universal fit
  • Free of detectable contaminants
  • Convenient, economical packaging
The universal fit PIPETMAN EXPERT Tips are designed to fit leading pipette brands, allowing you to stock one brand of tips saving time, money, and bench space. The tips are produced using state-of-the-art manufacturing techniques assuring lot-to-lot consistency and high performance. Volume graduations make it easy to visually check the volume aspirated. Color-codedMore Product Information