Ward's® Sterile, Disposable Cell Spreaders
Supplier: VWR International
470236-284 470236-286 470236-288 470236-290 470236-296
470236-284PK 188.35 CAD
470236-284 470236-286 470236-288 470236-290 470236-296
Ward's® Sterile, Disposable Cell Spreaders
Spreaders Cell Spreaders
Smooth-edged spreaders are excellent for spreading cultures and liquids without unwanted cutting, scraping, or gouging of agar during inoculation.
  • Available in multiple shapes and sizes
  • Smooth, rounded edges prevent damage to culture or media
  • Pre-sterilized, no flaming required
L-Shape: Ideal for spreading bacteriological cultures evenly. The shape allows the user to rotate and apply easily throughout an entire round plate or dish. They are made of ABS plastic.
T-Shape: Allows even pressure to be applied across the entire span of the 34 mmMore Product Information