Compact Refrigerated Category B Shipping Systems, Therapak®
Supplier: Therapak, LLC
380125G 380135G
76203-686CS 574.28 CAD
76203-686 76203-890
Compact Refrigerated Category B Shipping Systems, Therapak®
Mailers and Shippers
Compact refrigerated shippers are an ideal solution to meet the demands of medical and clinical specimen shipping.
  • Easy to use
  • Category B certified
  • Ship up to six 10 mL or smaller Blood Tubes
  • 95 kPa pressure rated secondary packaging
  • Complete shipping system
These certified Category B Shippers transport up to six 10 mL blood tubes. Precondition the included refrigerant packs for 24 hours at -20 ˚C and place in the cooler with the specimens. These shippers have been certified to transport Category B Specimens.
Certifications: Meets IATA Packing Instructions 650 requirements to transport Category B Specimens.