Ward's® Sterile Cell Strainers
Supplier: VWR International
C4010_WARDS C4040_WARDS C4070_WARDS C4100_WARDS C4015_WARDS C5100_WARDS
470236-274PK 97.95 CAD
470236-274 470236-276 470236-278 470236-280 470236-282 470236-310 470236-312 470236-314
Ward's® Sterile Cell Strainers
Cell Strainers
Ward's® Sterile Cell Strainers and Cell-Straining Kits provide all the necessary parts for filtration of cell suspensions.
  • Available as Kits (strainer, 50 ml tube, and cap) or as strainers alone
  • Easy to open under the hood, reducing contamination risk
Cell Straining Kits: Eliminate the contamination risks associated with opening multiple packages, and assembling separate pieces (i.e. tubes strainers and caps). The individual sterile packets mean users are no longer forced to expose entire bags of 50 ml tubes and corresponding caps in order to perform one filtration. Instead they can open a single packet directly under the hood.
Cell Strainers: If you don't need the accompanying tube that comes with the Cell Straining Kit, you can purchase 40 μm, 70 μm, or 100 μm cell strainers by themselves.