Dupont™ ProShield® 60 Coveralls with Laydown Collar and Elastic Wrists
Supplier: DuPont
NG125SWHSM002500 NG125SWHLG002500 NG125SWHXL002500 NG125SWH2X002500 NG125SWH4X002500 NG125SWH5X002500
CA13451-000CS 1044.4 CAD
CA13451-000 CA13451-002 CA13451-004 CA13451-006 CA13451-008 CA13451-010 CA13451-012 CA13451-014
Dupont™ ProShield® 60 Coveralls with Laydown Collar and Elastic Wrists
Disposable Garments
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Laydown collar
  • Elastic wrists and ankles
  • Serged seams
ProShield® 60 garments are made with a microporous film that is laminated to a spunbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation. Coveralls feature a laydown collar and elastic opening at wrists and ankles for a tighter fit.
Certifications: Thickness meets ASTM D751.