Biometra Thermal Cyclers, Analytik Jena
Supplier: Analytik Jena
846-070-231 846-4-070-215 846-070-232 846-070-233 846-4-070-251 846-070-230 846-070-271 846-070-220 846-4-070-311 846-4-070-723 846-070-261 846-2-070-301 846-070-234 846-4-070-724 846-2-070-301 846-4-070-211 846-4-070-720 846-4-070-723 846-070-231 846-4-070-214 846-4-070-210 846-4-070-241 846-4-070-202 846-070-235 846-4-070-311 846-4-070-201 846-4-070-214 846-070-221 846-070-232 846-2-070-311 846-4-070-212 846-070-220 846-4-070-301 846-4-070-224 846-070-261 846-070-224 846-4-070-724 846-070-222 846-4-070-215 846-4-070-280 846-070-221 846-4-070-241 846-070-224 846-4-070-224 846-070-234 846-4-070-720 846-2-070-311 846-4-070-212 846-070-271 846-4-070-201 846-4-070-202 846-4-070-213 846-4-070-200 846-070-230 846-4-070-251 846-4-070-200 846-4-070-213 846-070-222 846-070-235 846-4-070-210 846-070-233 846-4-070-301 846-4-070-211 846-4-070-280
CA76102-406EA 28473.84 CAD
CA76102-406 CA76102-404 76102-438 CA76102-408 CA75875-864 75873-972 75873-974 76102-440 75873-970 76102-444 76102-488 CA76102-452 76102-486 76102-442 76102-448 CA76102-456 CA76102-412 76102-446 CA76102-454 CA76102-410 CA76102-416 CA76102-458 CA76102-414 CA76102-418 76102-490 CA76102-460 CA75875-684 76102-450 76102-494 76102-492 CA76102-464 76102-498 CA76102-462 76102-496 CA76102-468 CA76102-466 CA76102-548 CA75875-688 CA75875-686 CA76102-470 75873-992 76102-422 CA76102-552 CA76102-474 76102-420 CA76102-550 CA76102-472 CA76102-556 76102-426 CA76102-478 76102-502 CA76102-554 76102-424 76102-500 CA76102-476 76102-506 76102-428 76102-504 76102-508 CA76102-480 76102-432 76102-430 76102-436 76102-434
Biometra Thermal Cyclers, Analytik Jena
Thermal Cyclers
The Biometra Thermal Cycler product family offers PCR devices ranging from standard to premium equipment – all designed in line with the common principles of being reliable, easy to use, and precise.
  • One block, two block, or 3 block systems available
  • Integrated 7" touchscreen control
  • Whisper quiet, with a low noise emission of 45 dB (max.)
  • High performance smart lid; Defined pressure control for highly reproducible results
  • Linear gradient tool allows easy gradient programming using the primer annealing temperature
  • Heated/Pressurized lid
  • Advanced user management allows individual rights settings for each user
  • Units include a two-year warranty
High-Speed Thermal Blocks: Biometra Thermal Cyclers are available in three different models, each with unique block formats. The Biometra TOne is a full-featured, budget friendly thermal cycler perfect for standard PCR work. The Biometra TAdvanced features twelve different block modules all with Quick Block Exchange allowing one thermal cycler to switch between various well combinations. These modules range from 60 well blocks to 384 well blocks as well as twin modules. For ultimate performance a 96 well block module made of silver (gold coated to prevent corrosion) is available. Twin modules feature two independently operating sample blocks. The Biometra TRIO provides three independent sample blocks which can run three different PCR reactions in parallel for maximum flexibility.
High-Performance Smart Lid (HPSL): The heated lid of the Biometra Thermal Cyclers are equipped with HPSL technology. An integrated slip clutch always maintains constant contact pressure, regardless of the shape and height of the plasticware. This optimizes the contact between the sample block well and the walls of the plasticware, resulting in reproducible conditions. As soon as the heated lid is closed, a rubber seal on the lid encapsulates the space surrounding the sample block. This closed space increases the sample block temperature uniformity and prevents condensation formation during the final PCR cooling step.
Biometra Thermal Cyclers come with a one-touch opening mechanism – just press the locking mechanism, and the heated lid swings open. A spring mechanism holds the lid in the open position, preventing it from dropping down. The locking mechanism automatically engages when the operator closes the lid.
Linear Gradient Tool (LGT) for maximum convenience:
Determining the optimum annealing temperature is a challenge when creating a new primer pair. Using the gradient tool allows the operator to find the optimum annealing temperature under experimental conditions and apply that temperature value to routine applications. Programming gradients usually means setting temperature for the first and the last row to determine the gradient range. For most thermal cyclers, the temperature interval is not the same from one row of the sample block to the next. In addition, the sigmoidal temperature curve of the gradient brings two disadvantages:
1. The automatically resulting temperatures for the other rows of the sample blocks, in particular in its center, are almost always odd-numbered.
2. The temperature differences between the rows are unevenly distributed, increasing towards the center of the sample block.
The Linear Gradient Tool enables programming of a temperature gradient with defined temperature intervals (increment) between the individual rows of the sample block. It allows the user to enter the calculated Tm value (minus e.g. 5 °C) and the desired temperature increment (e.g. ±1 °C) between the rows of the sample block. For maximum comfort, the Linear Gradient Tool now supports programming of even-numbered temperature values for a maximum number of rows.
State of the Art Touchscreen (Display): Biometra Thermal Cyclers feature a 7" state of the art color touchscreen user interface. The touchscreen is built-in at a flat angle to ensure reflection-free viewing and ergonomics. The software included makes for easy programming of PCR protocols through the use of Biometra’s proven spreadsheet philosophy and additionally a graphical programming mode. With one touch you can switch from spreadsheets to graphical programming making editing or creation of programs fast and easy.
Software: The Biometra Thermal Cyclers software offers a number of pre installed program templates for different applications. The program templates provide a general protocol structure and can be easily adapted for the current experiment. Editing a template or set-up of new protocols is quite simple due to the multi-step programming feature, which allows users to enter all of the parameters for every program step within a single screen. Thus, the multi-step programming renders constantly switching back and forth between different screens unnecessary and enables easy changes from step to step.