Disposable Sub-Micro Cuvettes, Type 700P, Firefly
Supplier: FIREFLYSCI, INC.
700PUV10A 700PUV10B
76016-594EA 402.39 CAD
76016-594 76016-596
Disposable Sub-Micro Cuvettes, Type 700P, Firefly
Cuvettes Spectrophotometer Cuvettes
These cuvettes are made from proprietary polycyclical olefin, and offer resistance to many aggressive solvents.
  • Fits most spectrophotometers without requiring adapters
  • Require only 70µL of sample volume
UV-transparent disposable cuvettes are useful for quick DNA, RNA, and protein assays in the 20–900nm range.
Packaging: Shipped 100 per box.