Containers, Wide Mouth, with Cap, WHEATON
Supplier: DWK Life Sciences (KIMBLE)
W209903 W209905 W209904
10811-456CS 394.73 CAD
10811-456 10813-208 10813-210
Containers, Wide Mouth, with Cap, WHEATON
Sample Containers General Purpose Containers
Durable PP container offers good chemical resistance.
  • Pre-attached white PP screw cap unlined or with foamed polyethylene liner
  • Linerless cap is autoclavable