DURAN® Erlenmeyer Flask, Wide Neck, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
212263903 212264402 212263603
76199-150CS 198.44 CAD
76199-150 76199-152 76199-742
DURAN® Erlenmeyer Flask, Wide Neck, DWK Life Sciences
Flasks Erlenmeyer Flasks
These wide neck erlenmeyer flasks are highly durable.
  • Easy-to-read white ceramic scale
  • Large labelling field for easy marking
  • Autoclavable at 121°C
Due to their conical form, flasks are ideally suited to the mixing of liquids. Uniform wall thickness facilitates heating applications. The wide neck enables easy filling and cleaning.
Certifications: DIN ISO 24450, USP Standard.